- Elaboration of simplified vinca alkaloids and phomopsin hybrids.
Elaboration of simplified vinca alkaloids and phomopsin hybrids.
Chemical biology & drug design (2010-07-28)
Quoc Anh Ngo, Fanny Roussi, Sylviane Thoret, Françoise Guéritte
PMID20659111
ABSTRAKT
Nine simplified vinca alkaloids and phomospin A hybrids, in which vindoline moiety has been replaced by a simpler scaffold, have been elaborated to evaluate their activity on the inhibition of tubulin polymerization. This article deals with the synthesis of various simplified vinca alkaloids, using a stereoselective coupling of catharantine with reactive aromatic compounds and methanol as well as their subsequent condensation with a large peptide chain mimicking those of phomopsin A. Biological evaluation and molecular modeling studies are also reported.