Merck
Neuromodulatory effect of GnRH on the synaptic transmission of the olfactory bulbar neural circuit in goldfish, Carassius auratus.
Journal of neurophysiology (2010-10-22)
Takafumi Kawai, Hideki Abe, Yasuhisa Akazome, Yoshitaka Oka
ABSTRAKT
Gonadotropin-releasing hormone (GnRH) is well known as a hypophysiotropic hormone that is produced in the hypothalamus and facilitates the release of gonadotropins from the pituitary gonadotropes. On the other hand, the functions of extrahypothalamic GnRH systems still remain elusive. Here we examined whether the activity of the olfactory bulbar neural circuits is modulated by GnRH that originates mainly from the terminal nerve (TN) GnRH system in goldfish (Carassius auratus). As the morphological basis, we first observed that goldfish TNs mainly express salmon GnRH (sGnRH) mRNA and that sGnRH-immunoreactive fibers are distributed in both the mitral and the granule cell layers. We then examined by extracellular recordings the effect of GnRH on the electrically evoked in vitro field potentials that arise from synaptic activities from mitral to granule cells. We found that GnRH enhances the amplitude of the field potentials. Furthermore, these effects were observed in both cases when the field potentials were evoked by stimulating either the lateral or the medial olfactory tract, conveying functionally different sensory information, separately, and suggesting that GnRH may modulate the responsiveness to wide categories of odorants in the olfactory bulb. Because GnRH also changed the paired-pulse ratio, it is suggested that the increased amplitude of the field potential results from changes in the presynaptic glutamate release of mitral cells rather than the increase in the glutamate receptor sensitivity of granule cells. These results suggest that TN regulates the olfactory responsiveness of animals appropriately by releasing sGnRH peptides in the olfactory bulbar neural circuits.