Merck
Efficacy and potential complications of difluprednate use for pediatric uveitis.
American journal of ophthalmology (2012-01-24)
Mark A Slabaugh, Erin Herlihy, Sharel Ongchin, Russell N van Gelder
ABSTRAKT
To evaluate the clinical effect of topical difluprednate in pediatric patients for treatment of noninfectious uveitis. Retrospective, observational case series. Twenty-six eyes of 14 pediatric patients with noninfectious uveitis who were treated with topical difluprednate were evaluated. Anterior and posterior cell grade, visual acuity, intraocular pressure (IOP), and cystoid macular edema (CME) were recorded at each visit. Main outcome measures were changes in anterior segment cell, CME, visual acuity, and IOP and development of a visually significant cataract. A significant (≥ 2-grade decrease or decrease to 0 in anterior segment cell) reduction in anterior segment inflammation was observed during treatment with topical difluprednate in 88% of eyes (22/25) when used as an adjuvant to systemic immunomodulatory therapy. In addition, improvement in CME associated with uveitis was seen in response to topical therapy with difluprednate in 78% of eyes with CME (7/9). A significant IOP response (IOP increase of ≥ 10 mm Hg from baseline and IOP ≥ 24 mm Hg) was seen in 50% of eyes (13/26) and in 50% of patients (7/14); 3 eyes of 2 patients required glaucoma surgery. Cataract formation or progression was observed in 39% of eyes (10/26) and in 43% of patients (6/14); 5 eyes of 3 patients required cataract surgery. Difluprednate is an effective agent for both control of anterior segment inflammation and reduction of CME in pediatric patients with uveitis when used as an adjuvant to systemic immunomodulatory therapy. A high rate of steroid-induced IOP elevation and cataract formation is seen in this population. Close monitoring of pediatric patients receiving difluprednate is recommended.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6α,9α-Difluoroprednisolone 21-acetate 17-butyrate, ≥98%