Merck
Enhanced Kα output of Ar and Kr using size optimized cluster target irradiated by high-contrast laser pulses.
Optics express (2012-01-26)
Lu Zhang, Li-Ming Chen, Da-Wei Yuan, Wen-Chao Yan, Zhao-Hua Wang, Cheng Liu, Zhong-Wei Shen, Anatoly Faenov, Tatiana Pikuz, Igor Skobelev, Vladimir Gasilov, Alexei Boldarev, Jing-Yi Mao, Yu-Tong Li, Quan-Li Dong, Xin Lu, Jing-Long Ma, Wei-Ming Wang, Zheng-Ming Sheng, Jie Zhang
ABSTRAKT
We observed that increasing the clusters size and laser pulse contrast can enhance the X-ray flux emitted by femtosecond-laser-driven-cluster plasma. By focusing a high contrast laser (10(-10)) on large argon clusters, high flux Kα-like X-rays (around 2.96 keV) is generated with a total flux of 2.5 × 10(11) photons/J in 4π and a conversion efficiency of 1.2 × 10-4. In the case of large Kr clusters, the best total flux for L-shell X-rays is 5.3 × 1011 photons/J with a conversion efficiency of 1.3 × 10-4 and, for the Kα X-ray (12.7 keV), it is 8 × 10(8) photons/J with a conversion efficiency of 1.6 × 10-6. Using this X-ray source, a single-shot high-performance X-ray imaging is demonstrated.