Merck
Oral alitretinoin in congenital ichthyosis: a pilot study shows variable effects and a risk of central hypothyroidism.
Acta dermato-venereologica (2012-01-27)
Agneta Gånemo, Mette Sommerlund, Anders Vahlquist
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
9-cis-Retinoic acid, ≥98% (HPLC)