Merck
  • New therapeutic strategy for amino acid medicine: prophylactic and healing promoting effect of monosodium glutamate against NSAID-induced enteropathy.
New therapeutic strategy for amino acid medicine: prophylactic and healing promoting effect of monosodium glutamate against NSAID-induced enteropathy.
Journal of pharmacological sciences (2012-02-02)
Kikuko Amagase, Akimu Ochi, Azusa Kojo, Ami Mizunoe, Masaya Taue, Naoya Kinoshita, Eiji Nakamura, Koji Takeuchi
ABSTRAKT
We reviewed the effect of monosodium glutamate (MSG) on the development and healing of nonsteroidal anti-inflammatory drug (NSAID)-induced small intestinal lesions in rats. Loxoprofen (60 mg/kg, p.o.) induced lesions in the small intestine within 24 h, accompanied by a decrease of Muc2 expression and an increase in enterobacterial invasion and inducible nitric oxide synthase (iNOS) expression. These lesions were prevented when MSG was given as a mixture of powdered food for 5 days before the loxoprofen treatment. This effect of MSG was accompanied by an increase in Muc2 expression / mucus secretion as well as the suppression of bacterial invasion and iNOS expression. These intestinal lesions healed spontaneously within 6 days, but the process was impaired by the repeated administration of low-dose loxoprofen (30 mg/kg) for 5 days after the ulceration, with the decrease of vascular endothelial derived growth factor (VEGF) expression and angiogenesis. The healing-impairing effect of loxoprofen was prevented by feeding 5% MSG for 5 days after the ulceration. These results suggest that MSG not only prevents loxoprofen-induced small intestinal damage but also promotes a healing of these lesions; the former is functionally associated with the increase in Muc2 expression / mucus secretion and the suppression of bacterial invasion and iNOS expression, while the latter is associated with the stimulation of VEGF expression/angiogenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Loxoprofen, solid