Merck
Chemical constituents of Euphorbia tangutica.
Natural product research (2012-03-20)
Ben-Yin Zhang, Huan Wang, Xiao-Dong Luo, Zhi-Zhi Du, Hai-Feng Wu, Jian-Wei Shen, Xiao-feng Zhang
ABSTRAKT
Sixteen known compounds isolated from the whole plants of Euphorbia tangutica, including phorbol-13-actate (1) previously synthesised and obtained from a natural source for the first time, were evaluated in vitro against a panel of human cancer cell lines using the MTT method. Among them, ergosterol (6) exhibited significant cytotoxic activity against HL-60 cell line with an IC(50) value of 3.3 µM, and 3β,5α-dihydroxy-15β-cinnamoyloxy-14-oxolathyra-6 Z,12 E-diene (7) also displayed moderate activity.