Merck
Salvianolic acid A protects human SH-SY5Y neuroblastoma cells against H₂O₂-induced injury by increasing stress tolerance ability.
Biochemical and biophysical research communications (2012-04-21)
Heng-ai Zhang, Mei Gao, Li Zhang, Yan Zhao, Li-li Shi, Bai-nian Chen, Yue-hua Wang, Shou-bao Wang, Guan-hua Du
ABSTRAKT
Salvianolic acid A (Sal A) is a polyphenol extracted from the root of the Salvia miltiorrhiza bunge. Hydrogen peroxide (H(2)O(2)) is a major reactive oxygen species (ROS), which has been implicated in stroke and other neurodegenerative diseases, including Alzheimer's disease and Parkinson's disease. In this study, we investigated the neuroprotective effects of Sal A in human SH-SY5Y neuroblastoma cells against H(2)O(2)-induced injury. Our results showed that cells pretreated with Sal A exhibited enhanced neuronal survival and that this protection was associated with an increase in adenosine triphosphate (ATP) and the stabilization of mitochondrial membrane potential. In addition, Sal A markedly decreased the excessive activation AMP-activated protein kinase (AMPK) and the serine-threonine protein kinase, Akt, in SH-SY5Ycells induced by H(2)O(2). In conclusion, our results demonstrated that Sal A protects SH-SY5Y cells against H(2)O(2)-induced oxidative stress and these protective effects are related to stress tolerance and not energy depletion via inhibition of the AMPK and Akt signaling pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Salvianolic acid A, analytical standard