- Lack of detectable metabolism for solubilized 2,3,4-trimethylpentane by rat kidney proximal tubules.
Lack of detectable metabolism for solubilized 2,3,4-trimethylpentane by rat kidney proximal tubules.
Toxicology letters (1990-12-01)
N J DelRaso, D R Mattie, C S Godin
PMID2260128
ABSTRAKT
Primary proximal tubule suspension cultures exposed to solubilized 2,3,4-trimethylpentane (2,3,4-TMP) resulted in a linear dose response, as determined by cellular lactate dehydrogenase leakage. The EC50 for 2,3,4-TMP was 16.3 mM. Metabolite analysis by gas chromatography/mass spectrometry of supernate and cell extracts from cultures exposed to 2,3,4-TMP (12.0 mM) failed to detect the presence of metabolites. Electron-microscopic examination of proximal tubules exposed to 2,3,4-TMP indicated ultrastructural changes that included increased mitochondrial swelling, increased vesiculation, decreased microvilli and pyknotic nuclei. This study indicates that kidney proximal tubules do not appear to metabolize 2,3,4-TMP.