Merck
[Stages of bone regeneration and foundation of pharmacоlogical correction of the mandible reparative osteogenesis].
Stomatologiia (2012-06-09)
M B Shvyrkov
ABSTRAKT
Influents of some medicines on the mandible reparative regeneration in experimental investigations are present in the manuscript. It was discovered that application of parathyroid hormone (PTG) in first 3 days after the mandible fracture decreases of quantity complications on 30% and contributes strong consolidation of bone fragments. Metiluracil in the same period increases complications quantity on 36% and bone fragments consolidation strength was for sure less than after PTG. It was found PTG and vitamin D3 in first resorbtion stage accelerate bone consolidation, retabolil (nandrolone), dexametasone, testosterone and vitamin E slow down bone consolidation. Role of vitamin A, indometazin and calcitonin was doubtful. Practical doctors and experimenters co-ordination efforts are necessary for elaboration of new tactic and strategy of patient treatment with bone trauma.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6-Methyluracil, 97%
Nandrolone decanoate for system suitability, European Pharmacopoeia (EP) Reference Standard
Nandrolone decanoate for peak identification, European Pharmacopoeia (EP) Reference Standard
Nandrolone decanoate, European Pharmacopoeia (EP) Reference Standard