Merck
  • [Oral combined hormonal contraception containing nomegestrol acetate and 17beta-oestradiol--the statement of Polish Gynecological Society Experts Group].
[Oral combined hormonal contraception containing nomegestrol acetate and 17beta-oestradiol--the statement of Polish Gynecological Society Experts Group].
Ginekologia polska (2012-06-21)
Romuald Debski, Jan Kotarski, Tomasz Paszkowski, Leszek Pawelczyk, Ryszard Poreba, Violetta Skrzypulec-Plinta, Jacek Tomaszewski
ABSTRAKT
Nomegestrol acetate (NOMAC) combined with E2 (Zoely) is a monophasic oral contraceptive (OC) which safety and efficacy was confirmed in a number of level I evidence clinical trials. Zoely is highly effective OC, especially in overweight and obese patients, with good cycle control, safe and well tolerated. NOMAC/E2 combination causes no or minimal weight gain and is characterized by minimal influence on bone mineral density or blood pressure and presence of acne. Moreover lipids profile, carbohydrates metabolism, haemostasis and endocrine glands functioning were not affected. High tolerance and acceptance of NOMAC/E2 combination by women, low adverse event profile, fast recovery of ovarian activity and ovulation is a reasonable treatment tool in everyday practice.