Merck
Sufentanil postconditioning protects the myocardium from ischemia-reperfusion via PI3K/Akt-GSK-3β pathway.
The Journal of surgical research (2012-06-26)
Qiao-ling Wu, Tu Shen, Hong Ma, Jun-ke Wang
ABSTRAKT
Previous studies have shown that opioid postconditioning reduces apoptosis through antiapoptotic signaling. The present study evaluated whether sufentanil could induce cardioprotection after ischemia-reperfusion (I/R) and whether the PI3K/Akt-GSK-3β pathway modulates antiapoptotic proteins in sufentanil postconditioning. We subjected male Sprague-Dawley rats to 30 min of myocardial ischemia and 2 h of reperfusion. We randomized rats into seven groups: sham, I/R, sufentanil postconditioning (I/R+sufen), sham plus sufentanil (sham+sufen), sham plus 15 μg · kg(-1) intravenous wortmannin (PI3K inhibitor), I/R plus wortmannin, and sufentanil plus wortmannin. We induced sufentanil postconditioning with 3 μg · kg(-1) sufentanil for 3 min in the beginning of reperfusion after 30 min ischemia. We assessed hemodynamics, myocardial infarct size, number of apoptotic cardiomyocytes, total Akt and GSK-3β, phosphorylated Akt and GSK-3β, caspase-3, Bax, and Bcl-2 protein expression. The I/R+sufen group had significantly reduced infarct size compared with the I/R group (23.3% ± 9.0% versus 50.1% ± 7.4%; P < 0.05). The apoptotic index of cardiomyocytes was significantly reduced with sufentanil treatment (20.0% ± 3.5%) compared with the I/R group (47.0% ± 6.3%; P < 0.05). The I/R+sufen group reduced the expression of protein-cleaved caspase-3 and Bax, and increased Bcl-2, phosphorylated Akt, and GSK3β compared with the I/R group. Wortmannin eliminated the cardioprotection produced with sufentanil treatment. Sufentanil postconditioning can induce myocardial protection by activating the PI3K/Akt-GSK-3β pathway and modulating Bax and Bcl-2 expression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sufentanil Citrate solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Sufentanil-D5 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®