Merck
  • A comparative 'bottom up' proteomics strategy for the site-specific identification and quantification of protein modifications by electrophilic lipids.
A comparative 'bottom up' proteomics strategy for the site-specific identification and quantification of protein modifications by electrophilic lipids.
Journal of proteomics (2012-07-31)
Bingnan Han, Michael Hare, Samanthi Wickramasekara, Yi Fang, Claudia S Maier
ABSTRAKT
We report a mass spectrometry-based comparative "bottom up" proteomics approach that combines d(0)/d(4)-succinic anhydride labeling with commercially available hydrazine (Hz)-functionalized beads (Affi-gel Hz beads) for detection, identification and relative quantification of site-specific oxylipid modifications in biological matrices. We evaluated and applied this robust and simple method for the quantitative analysis of oxylipid protein conjugates in cardiac mitochondrial proteome samples isolated from 3- and 24-month-old rat hearts. The use of d(0)/d(4)-succinic anhydride labeling, Hz-bead based affinity enrichment, nanoLC fractionation and MALDI-ToF/ToF tandem mass spectrometry yielded relative quantification of oxylipid conjugates with residue-specific modification information. Conjugation of acrolein (ACR), 4-hydroxy-2-hexenal (HHE), 4-hydroxy-2-nonenal (HNE) and 4-oxo-2-noneal (ONE) to cysteine, histidine and lysine residues were identified. HHE conjugates were the predominant subset of Michael-type adducts detected in this study. The HHE conjugates showed higher levels in mitochondrial preparations from young heart congruent with previous findings by others that the n-3/n-6 PUFA ratio is higher in young heart mitochondrial membranes. Although this study focuses on protein adducts of reactive oxylipids, the method might be equally applicable to protein carbonyl modifications caused by metal catalyzed oxidation reactions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)