Merck
  • Assessment of salivary free cortisol levels by liquid chromatography with tandem mass spectrometry (LC-MS/MS) in patients treated with mitotane.
Assessment of salivary free cortisol levels by liquid chromatography with tandem mass spectrometry (LC-MS/MS) in patients treated with mitotane.
Hormones (Athens, Greece) (2012-08-22)
Cinzia Carrozza, Rosa Lapolla, Jacopo Gervasoni, Carlo Antonio Rota, Pietro Locantore, Alfredo Pontecorvi, Cecilia Zuppi, Silvia Persichilli
ABSTRAKT
Mitotane is an adrenocytolytic agent used in adrenocortical carcinoma, inducing adrenal insufficiency, requiring replacement treatment. Such therapy is not easy to monitor because of mitotane interference. Salivary cortisol reflects a free fraction of plasma cortisol and may be useful in such patients. The aim of our study was to evaluate salivary cortisol by HPLC coupled to tandem-mass spectrometry (LC-MS/MS) and by an electrochemiluminescence immunoassay (ECLIA) in patients treated with mitotane. We enrolled 6 patients receiving mitotane and 2 Addison disease patients as negative controls and determined salivary cortisol rhythm. We also determined the salivary cortisol rhythm in 8 healthy subjects. Salivary samples (n=112) were assayed by ECLIA, using Roche Modular E170, and by LC-MS/MS. The mean values obtained by ECLIA were significantly higher than those obtained by LC-MS/MS in the mitotane group (p<0.001). In fact, in the group measured by LC-MS/MS, we observed several peaks eluting at a retention time different from the cortisol group, presumably due to cortisol-like analogues. In Addison disease, since steroidogenesis is absent, salivary cortisol values measured by the two methods did not show any significant difference (p=0.61). Salivary cortisol measured by LC-MS/MS is a selective method, excluding cortisol analogues accumulating in treated patients. Therefore, LC-MS/MS offers an effective system to monitor replacement therapy in mitotane treated patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
2,4′-DDD, PESTANAL®, analytical standard