Merck
  • Glycolaldehyde impairs neutrophil biochemical parameters by an oxidative and calcium-dependent mechanism--protective role of antioxidants astaxanthin and vitamin C.
Glycolaldehyde impairs neutrophil biochemical parameters by an oxidative and calcium-dependent mechanism--protective role of antioxidants astaxanthin and vitamin C.
Diabetes research and clinical practice (2012-08-28)
Beatriz Alves Guerra, Anaysa Paola Bolin, Ana Carolina Morandi, Rosemari Otton
ABSTRAKT
The present study examined the effects of glycolaldehyde (GC) on biochemical parameters of human neutrophils and whether the antioxidant astaxanthin associated with vitamin C can modulate these parameters. Neutrophils from healthy subjects were treated with GC (1mM) followed or not by the antioxidants astaxanthin (2 μM) and vitamin C (100 μM). We examined the phagocytic capacity, hypochlorous acid, myeloperoxidase (MPO) and glucose-6-phosphate dehydrogenase (G6PDH) activities, cytokines and [Ca(2+)](i). Also, superoxide anion, hydrogen peroxide, nitric oxide production, antioxidant enzyme activities and glutathione-recycling system were evaluated. GC promoted a marked reduction on the phagocytic capacity, maximal G6PDH and MPO activities, hypochlorous acid production and release of IL-1β, IL-6 and TNF-α cytokines. Some impairment in the neutrophils biochemical parameters appears to be mediated by oxidative stress through ROS/RNS production and calcium reduction. Oxidative stress was evidenced by reduction in the activities of the main antioxidant enzymes, GSH/GSSG ratio and in the increment of O(2)(-) and H(2)O(2) and NO. Treatment of cells with the combination of the antioxidants astaxanthin and vitamin C was able to restore some neutrophils function mainly by decreasing ROS/RNS production and improving the redox state. Overall, our findings demonstrate that GC modulates several neutrophils biochemical parameters in vitro.