Merck
Nomegestrol acetate/estradiol: in oral contraception.
Drugs (2012-09-07)
Lily P H Yang, Greg L Plosker
ABSTRAKT
Nomegestrol acetate/estradiol is a combined oral contraceptive with approval in many countries. This fixed-dose combination tablet contains nomegestrol acetate, a highly selective progestogen, and estradiol, a natural estrogen. It is the first monophasic combined oral contraceptive to contain estradiol, and is taken in 28-day cycles, consisting of 24 active therapy days with 4 placebo days (i.e. 24/4-day cycles). In two large, 1-year, randomized, open-label, multicentre, phase III trials in healthy adult women (aged 18-50 years), nomegestrol acetate/estradiol was at least as effective as drospirenone/ethinylestradiol as contraceptive therapy, as the pregnancy rates in women aged 18-35 years (primary efficacy population) in terms of the Pearl Index (primary endpoint) were numerically lower with nomegestrol acetate/estradiol, although the between-group difference was not statistically significant. In both trials, nomegestrol acetate/estradiol was given in a 24/4-day cycle, and drospirenone/ethinylestradiol was given in a 21/7-day cycle. The criteria for using condoms in case of forgotten doses were less stringent in the nomegestrol acetate/estradiol group than in the drospirenone/ethinylestradiol group. Nomegestrol acetate/estradiol therapy for up to 1 year was generally well tolerated in healthy adult women, with an acceptable tolerability profile in line with that expected for a combined oral contraceptive. The most commonly reported adverse events were acne and abnormal withdrawal bleeding (most often shorter, lighter or absent periods). Overall, compared with drospirenone/ethinylestradiol, nomegestrol acetate/estradiol appeared to be associated with less favourable acne-related outcomes, and shorter, lighter or absent periods.