Merck
Hair ethyl glucuronide is highly sensitive and specific for detecting moderate-to-heavy drinking in patients with liver disease.
Alcohol and alcoholism (Oxford, Oxfordshire) (2012-09-28)
Scott H Stewart, David G Koch, Ira R Willner, Patrick K Randall, Adrian Reuben
ABSTRAKT
Hair ethyl glucuronide (EtG) is a promising biomarker of moderate-to-heavy alcohol consumption and may have utility in detecting and monitoring alcohol use in clinical populations where alcohol use is of particular importance. This study evaluated the relationship between hair EtG and drinking in patients with liver disease. The subjects (n = 200) were patients with liver disease who presented for care at a university medical center. Alcohol use during the 3 months preceding participation in the study was assessed, and a sample of hair was obtained for EtG testing. Classification of drinking status (any drinking or averaging at least 28 g per day) by hair EtG was evaluated, as well as the effects of liver disease severity and demographic and hair care factors. The area under the receiver operating characteristic curve for detecting an average of 28 g or more per day during the prior 90 days was 0.93. The corresponding sensitivity and specificity of hair EtG ≥8 pg/mg for averaging at least 28 g of ethanol per day were 92 and 87%, respectively. Cirrhosis and gender may have a modest influence on the relationship between drinking and hair EtG. Hair EtG was highly accurate in differentiating subjects with liver disease averaging at least 28 g of ethanol per day from abstainers and lighter drinkers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Ethyl-β-D-glucuronide, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethyl-β-D-glucuronide, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®