Merck
Xenon-enhanced dual-energy CT of patients with asthma: dynamic ventilation changes after methacholine and salbutamol inhalation.
AJR. American journal of roentgenology (2012-10-26)
Won Wha Kim, Chang Hyun Lee, Jin Mo Goo, Sang Joon Park, Jong Hyo Kim, Eun-Ah Park, Sang-Heon Cho
ABSTRAKT
The purpose of this study was to evaluate the use of xenon-enhanced dual-energy CT of the chest to assess ventilation changes after methacholine and salbutamol inhalation in subjects with asthma and healthy subjects. Twenty-five subjects with asthma and 10 healthy subjects underwent three-phase (basal, after methacholine inhalation, after salbutamol inhalation) xenon-enhanced chest CT. Each phase was composed of wash-in and washout scans. For visual analysis, two radiologists evaluated ventilation defects and gas trapping lobe by lobe on a 10-point scale. Total ventilation defect and gas trapping scores were calculated by adding ventilation defect and gas trapping scores. Xenon and total lung volume were quantified automatically. Total xenon concentration index was calculated as total xenon concentration divided by lung volume. Repeated measures analysis of variance and Student t test were used for comparisons of total ventilation defect score, total gas trapping score, and total xenon concentration index between the two groups. The Friedman test was used for within-group analysis. In the basal state, subjects with asthma had a higher total ventilation defect score (p = 0.004) and higher total gas trapping score (p = 0.05) than did healthy subjects. On washout images, total ventilation defect score, total gas trapping score, and total xenon concentration index after methacholine and salbutamol inhalation were statistically different between the two groups (p < 0.05). However, total xenon concentration index on wash-in images was not significantly different between the two groups. In within-group analysis, total ventilation defect score and total gas trapping score in subjects with asthma and total ventilation defect score in healthy subjects increased significantly after methacholine inhalation and decreased significantly after salbutamol inhalation (p < 0.05). Xenon-enhanced chest CT may be a useful technique for visualizing dynamic changes in airflow in response to methacholine and salbutamol inhalation in patients with asthma. Optimization of the protocol for radiation exposure is warranted.