- The rare inherited coagulation disorders.
The rare inherited coagulation disorders.
Pediatric blood & cancer (2012-10-31)
Paula H B Bolton-Maggs
PMID23109366
ABSTRAKT
The rare inherited coagulation disorders (RICD) are uncommon and thus not well-defined in terms of severity or management. Inheritance is autosomal; in some of these disorders in the heterozygote state affected individuals may be mildly symptomatic. Severe deficiencies are more common in association with consanguinity. Factor X and factor XIII deficiency have the most severe manifestations, while factor XI deficiency is the least severe. Factor VII and factor XI deficiencies show a poor relationship between the factor level and bleeding risk. Unlike hemophilia, women are equally affected by these RICD and can have problems related to menstruation and childbirth.