Merck
Evaluation of organochlorine contamination in Indo-Pacific humpback dolphins (Sousa chinensis) from the Pearl River Estuary, China.
The Science of the total environment (2013-01-03)
Yinglin Wu, Jingchun Shi, Gene J Zheng, Ping Li, Bo Liang, Tufeng Chen, Yuping Wu, Wenhua Liu
ABSTRAKT
Organochlorine compounds, including polychlorinated biphenyls (PCBs), dichlorodiphenyl trichloroethane (DDTs), hexachlorocyclohexane (HCHs), chlordanes (CHLs) and hexachlorobenzene (HCB) were analyzed in blubber samples collected from fifteen Indo-Pacific humpback dolphins (Sousa chinensis) stranded in the Pearl River Delta during the period from 2004 to 2009. In general, the magnitude of the mean concentrations is: ∑DDTs>∑PCBs>∑HCHs>∑CHLs>HCB. DDT levels found in this study were very high, with the ΣDDT ranging from 845 to 179,000 ng/g wet weight. Of particular importance is the fact that new inputs from diverse sources still exist in the Pearl River Delta. ∑PCBs ranged from 123 to 10,200 ng/g wet weight. More studies are necessary for a better understanding of these organochlorine contaminants and their impact on the conservation of humpback dolphins residing in the Pearl River Estuary waters.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
4,4′-DDT, PESTANAL®, analytical standard
Supelco
Hexachlorobenzene solution, certified reference material, 1000 μg/mL in acetone
Supelco
Lindane, PESTANAL®, analytical standard
Supelco
Lindane, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hexachlorobenzene, PESTANAL®, analytical standard
Supelco
4,4′-DDT solution, certified reference material, 5000 μg/mL in methanol