Merck
High age and low sodium urine concentration are associated with poor survival in patients with hepatorenal syndrome.
Annals of hepatology (2013-01-08)
Matthias Hinz, Alexander Wree, Christoph Jochum, Lars P Bechmann, Fuat Saner, Alexander L Gerbes, Guido Gerken, Ali Canbay
ABSTRAKT
Combination therapy with terlipressin and albumin substitution is considered a widely accepted treatment regimen for patients with hepatorenal syndrome (HRS). However, only half of the patients respond to treatment and to date albumin substitution and terlipressin therapy are among the most expensive medical treatments available for patients with liver diseases. Thus, we aimed to identify clinical and etiological parameters to predict treatment response and overall mortality in patients with HRS. We retrospectively evaluated 21 patients, 13 male/8 female, aged 43-72 years with HRS. Four patients were transplanted after following combination treatment. Terlipressin was administered by continuous intravenous perfusion (2-6 mg/d) and albumin drips (50 mg) were given daily. Treatment response was defined by a decrease in serum creatinine level to ≤ 1.5 mg/dL or by a ≥ 50% reduction of the baseline concentration. 57% of the patients responded to treatment, which was associated with improved survival at day 60, compared to non-responders. However, the overall mortality was not different between the two groups. Median age of 63 years was a significant negative predictor for therapy response. High baseline urinary sodium levels were of prognostic value for survival. The Model of End stage Liver Disease score (MELD score) did not correlate with therapy response. In conclusion high age is a predictor of non-response. Low urinary sodium before treatment is associated with poor survival. Terlipressin and albumin co-treatment is associated with increased two-months survival rate. This seemingly moderate extension in survival rate can, however, be decisive for obtaining liver transplantation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
[Lys8]-Vasopressin, ≥98% (HPLC)