Merck
Mutational analysis of TAC3 and TACR3 genes in patients with idiopathic central pubertal disorders.
Arquivos brasileiros de endocrinologia e metabologia (2013-01-19)
Cintia Tusset, Sekoni D Noel, Ericka B Trarbach, Letícia F G Silveira, Alexander A L Jorge, Vinicius N Brito, Priscila Cukier, Stephanie B Seminara, Berenice B de Mendonça, Ursula B Kaiser, Ana Claudia Latronico
ABSTRAKT
To investigate the presence of variants in the TAC3 and TACR3 genes, which encode NKB and its receptor (NK3R), respectively, in a large cohort of patients with idiopathic central pubertal disorders. Two hundred and thirty seven patients were studied: 114 with central precocious puberty (CPP), 73 with normosmic isolated hypogonadotropic hypogonadism (IHH), and 50 with constitutional delay of growth and puberty (CDGP). The control group consisted of 150 Brazilian individuals with normal pubertal development. Genomic DNA was extracted from peripheral blood and the entire coding region of both TAC3 and TACR3 genes were amplified and automatically sequenced. We identified one variant (p.A63P) in NKB and four variants, p.G18D, p.L58L (c.172C>T), p.W275* and p.A449S in NK3R, which were absent in the control group. The p.A63P variant was identified in a girl with CPP, and p.A449S in a girl with CDGP. The known p.G18D, p.L58L, and p.W275* variants were identified in three unrelated males with normosmic IHH. Rare variants in the TAC3 and TACR3 genes were identified in patients with central pubertal disorders. Loss-of-function variants of TACR3 were associated with the normosmic IHH phenotype.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Neurokinin B, ≥95% (HPLC)