Merck
Long-term safety and efficacy of a pasteurized nanofiltrated prothrombin complex concentrate (Beriplex P/N): a pharmacovigilance study.
British journal of anaesthesia (2013-01-22)
A A Hanke, C Joch, K Görlinger
ABSTRAKT
The rapid reversal of the effects of vitamin K antagonists is often required in cases of emergency surgery and life-threatening bleeding, or during bleeding associated with high morbidity and mortality such as intracranial haemorrhage. Increasingly, four-factor prothrombin complex concentrates (PCCs) containing high and well-balanced concentrations of vitamin K-dependent coagulation factors are recommended for emergency oral anticoagulation reversal. Both the safety and efficacy of such products are currently in focus, and their administration is now expanding into the critical care setting for the treatment of life-threatening bleeding and coagulopathy resulting either perioperatively or in cases of acute trauma. After 15 yr of clinical use, findings of a pharmacovigilance report (February 1996-March 2012) relating to the four-factor PCC Beriplex P/N (CSL Behring, Marburg, Germany) were analysed and are presented here. Furthermore, a review of the literature with regard to the efficacy and safety of four-factor PCCs was performed. Since receiving marketing authorization (February 21, 1996), ~647 250 standard applications of Beriplex P/N have taken place. During this time, 21 thromboembolic events judged to be possibly related to Beriplex P/N administration have been reported, while no incidences of viral transmission or heparin-induced thrombocytopenia were documented. The low risk of thromboembolic events reported during the observation period (one in ~31 000) is in line with the incidence observed with other four-factor PCCs. In general, four-factor PCCs have proven to be well tolerated and highly effective in the rapid reversal of vitamin K antagonists.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Factor IX human, aqueous glycerol solution, ≥145 units/mg protein