- [Gestational diabetes insipidus during a twin pregnancy].
[Gestational diabetes insipidus during a twin pregnancy].
Annales francaises d'anesthesie et de reanimation (2013-02-06)
M De Mesmay, A Rigouzzo, T Bui, N Louvet, I Constant
PMID23380272
ABSTRAKT
Gestational diabetes insipidus is an uncommon clinical disease whose prevalence is approximately two to three pregnancies per 100,000. It may be isolated or associated with preeclampsia. We report a case of gestational diabetes insipidus in a twin pregnancy, originally isolated during two months, and secondarily complicated by HELLP-syndrome. We recall the specific pathophysiology of polyuric-polydipsic syndrome during pregnancy and summarize its various causes. Finally, we discuss the indications, in case of isolated gestational diabetes insipidus, of treatment by dDAVP.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Desmopressin, European Pharmacopoeia (EP) Reference Standard