Merck
  • Capillary electrophoresis with laser-induced fluorescence detection of proteins from two types of complex sample matrices: food and biological fluids.
Capillary electrophoresis with laser-induced fluorescence detection of proteins from two types of complex sample matrices: food and biological fluids.
Methods in molecular biology (Clifton, N.J.) (2013-02-07)
Raul Garrido-Medina, Angel Puerta, Cristina Pelaez-Lorenzo, Zuly Rivera-Monroy, Andras Guttman, Jose Carlos Diez-Masa, Mercedes de Frutos
ABSTRAKT
Sample preparation and laser-induced fluorescence detection are two key steps of the analytical methodology to determine by capillary electrophoresis low concentrations of proteins in complex sample matrices. In this chapter the options of performing both steps in different ways are shown by detailing the analysis of the allergen β-lactoglobulin in food products for infants and the analysis of the isoforms of alpha 1-acid glycoprotein, a potential biomarker, in serum and secretome.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-(2-Furoyl)quinoline-2-carboxaldehyde, ≥95% (HPLC), suitable for fluorescence, BioReagent