Merck
  • Protein kinase Cα inhibitor protects against downregulation of claudin-1 during epithelial-mesenchymal transition of pancreatic cancer.
Protein kinase Cα inhibitor protects against downregulation of claudin-1 during epithelial-mesenchymal transition of pancreatic cancer.
Carcinogenesis (2013-02-08)
Daisuke Kyuno, Takashi Kojima, Hiroshi Yamaguchi, Tatsuya Ito, Yasutoshi Kimura, Masafumi Imamura, Akira Takasawa, Masaki Murata, Satoshi Tanaka, Koichi Hirata, Norimasa Sawada
ABSTRAKT
Protein kinase Cα (PKCα) is highly expressed in pancreatic cancer. However, the effects of PKCα on Snail and claudin-1, which play crucial roles in epithelial cell polarity during epithelial-mesenchymal transition (EMT), remain unclear. In this study, we investigated the mechanisms of regulation of Snail and claudin-1 via a PKCα signal pathway during EMT in pancreatic cancer cells and in normal human pancreatic duct epithelial cells (HPDEs). By immunostaining, overexpression of PKCα and downregulation of claudin-1 were observed in poorly differentiated human pancreatic cancer tissues and the pancreatic cancer cell line PANC-1. Treatment with the PKCα inhibitor Gö6976 transcriptionally decreased Snail and increased claudin-1 in PANC-1 cells. The PKCα inhibitor prevented upregulation of Snail and downregulation of claudin-1 during EMT induced by transforming growth factor-β1 (TGF-β1) treatment and under hypoxia in PANC-1 cells. The effects of the PKCα inhibitor were in part regulated via an extracellular signal-regulated kinase (ERK) signaling pathway. The PKCα inhibitor also prevented downregulation of the barrier function and fence function during EMT in well-differentiated pancreatic cancer cell line HPAC. In normal HPDEs, the PKCα inhibitor transcriptionally induced not only claudin-1 but also claudin-4, -7 and occludin without a change of Snail. Treatment with the PKCα inhibitor in normal HPDEs prevented downregulation of claudin-1 and occludin by TGF-β1 treatment and enhanced upregulation of claudin-1, -4, -7 and occludin under hypoxia. These findings suggest that PKCα regulates claudin-1 via Snail- and mitogen-activated protein kinase/ERK-dependent pathways during EMT in pancreatic cancer. Thus, PKCα inhibitors may be potential therapeutic agents against the malignancy of human pancreatic cancer cells.