Merck
Jujube honey from China: physicochemical characteristics and mineral contents.
Journal of food science (2013-03-06)
Juan Zhou, Zhirong Suo, Pinpin Zhao, Ni Cheng, Hui Gao, Jing Zhao, Wei Cao
ABSTRAKT
We investigated and compared the physicochemical properties (moisture, color, ash, pH, electrical conductivity, free acidity, lactonic acidity, total acidity, fructose, glucose, sucrose, diastase activity, and HMF) and mineral contents (Al, Ca, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Na, Ni, As, Cd, Pb, and Zn), as well as total proline and total protein contents of 23 jujube honey samples collected from different regions of China. The mineral content was determined by inductively coupled plasma-optical emission spectrometry (ICP-OES). The physicochemical values were in the range of approved limits (conforming to EU legislation) in all 23 samples. The physicochemical properties of jujube honey showed significant variations among samples. The mean pH value of the jujube honeys was 6.71. The most abundant minerals were potassium, calcium, sodium, and magnesium, ranging between 1081.4 and 2642.9, 97.1 and 194.2, 7.79 and 127.8, and 10.36 and 24.67 mg/kg, respectively, and potassium made up 71% of the total mineral content. This study demonstrated remarkable variation in physicochemical parameters and mineral contents of jujube honey, mainly depending on its geographic source.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
5-(Hydroxymethyl)furfural, analytical standard
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Furfural, ≥98%, FCC, FG
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%
Sigma-Aldrich
5-(Hydroxymethyl)furfural, ≥99%, FG