Merck
Butanolides from methanolic extract of Litsea glutinosa.
Chemistry & biodiversity (2013-03-16)
Nisha Agrawal, Deepika Pareek, Sonal Dobhal, Mahesh C Sharma, Yogesh C Joshi, Mahabeer P Dobhal
ABSTRAKT
Phytochemical investigations of a MeOH extract obtained from the heartwoods of the Litsea glutinosa (Lauraceae) led to the isolation and characterization of four new butenolides, (3R,4S,5S)-2-hexadecyl-3-hydroxy-4-methylbutanolide 1, litsealactone C (2), and litsealactone D (4), litsealactone G (5), and a new benzoic acid derivative named eusmoside C (3). The structures of these compounds were elucidated on the basis of spectral studies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
Sigma-Aldrich
2(5H)-Furanone, 98%
Sigma-Aldrich
γ-Butyrolactone, ReagentPlus®, ≥99%