Merck
Nitric oxide suppresses L-type calcium currents in basilar artery smooth muscle cells in rabbits.
Neurological research (2013-04-02)
Naveen Sharma, Janardhan Prasad Bhattarai, Pyoung Han Hwang, Seong Kyu Han
ABSTRAKT
Nitric oxide (NO) is well known to be a vasodilator, and NO donor compounds are currently used for treating vasospasm following subarachnoid hemorrhage. However, the action mechanism of cerebral vascular relaxation is not yet clear. L-type calcium channels have been determined to play an essential role in smooth muscle contraction. To investigate the role of L-type calcium channels in NO-induced relaxation of basilar smooth muscle cells, we examined the effect of the NO donor, sodium nitroprusside (SNP) on calcium (Ca2+) currents using smooth muscle cells isolated from a rabbit basilar artery. The smooth muscle cells were isolated from rabbit basilar artery by enzyme treatment. To identify L-type Ca2+ currents, we used cesium chloride, a potassium channel blocker and Bay K8644, an activator of L-type Ca2+ channel. The L-type calcium currents (91±13.0 pA; n = 11) were significantly reduced by SNP (32±5 pA; n = 11; P<0.05). 1H-[1,2,4] Oxadiazolo [4,3-a] quinoxalin-1-one, a 3',5'-cyclic guanosine monophosphate inhibitor, blocked the effect of SNP on L-type Ca2+ currents, and similar results were obtained after the application of 7-nitroindazole, a specific NO synthase inhibitor. Furthermore, inward currents were enhanced by Bay K8644 (170±22 pA; n = 5) and were suppressed by SNP (54±13 pA; n = 5; P<0.05). These results demonstrate that NO suppresses the L-type Ca2+ currents in rabbit basilar smooth muscle cells, and suggest that L-type Ca2+ channels may play a pivotal role in NO-induced vascular relaxation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium nitroferricyanide(III) dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium nitroprusside dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%