Merck
Investigation of a planar optical waveguide in 2D PPLN using helium implantation technique.
Optics express (2013-04-03)
Q Ripault, M W Lee, F Mériche, T Touam, B Courtois, E Ntsoenzok, L-H Peng, A Fischer, A Boudrioua
ABSTRAKT
In this work, we report the investigation of a planar waveguide in a 2D periodically-poled lithium niobate (PPLN). The waveguide is fabricated by helium (He(+)) implantation at 2 MeV and a fluence of 1.5 x 10(16) ions/cm(2). Second harmonic generation (SHG) at 532 nm using a Q-switched laser and a CW laser diode at 1064 nm, was measured as a function of angular distribution and temperature. The experimental results show higher gain in SHG conversion efficiency in the waveguide than in the bulk 2D PPLN. In particular, SHGs from 2D reciprocal lattice vectors (RLV) are observed and studied.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lithium niobate, 99.9% trace metals basis
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis
Niobium, IRMM®, certified reference material, 0.02 mm foil
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil
Niobium, IRMM®, certified reference material, 0.5 mm wire