Merck
Treatment of refractory infectious keratitis with corneal collagen cross-linking window absorption.
Cornea (2013-04-11)
Riccardo Vinciguerra, Pietro Rosetta, Mario R Romano, Claudio Azzolini, Paolo Vinciguerra
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material