Merck
The effect of exogenous GLP-1 on food intake is lost in male truncally vagotomized subjects with pyloroplasty.
American journal of physiology. Gastrointestinal and liver physiology (2013-04-20)
Astrid Plamboeck, Simon Veedfald, Carolyn F Deacon, Bolette Hartmann, André Wettergren, Lars B Svendsen, Søren Meisner, Claus Hovendal, Tina Vilsbøll, Filip K Knop, Jens J Holst
ABSTRAKT
Rapid degradation of glucagon-like peptide-1 (GLP-1) by dipeptidyl peptidase-4 suggests that endogenous GLP-1 may act locally before being degraded. Signaling via the vagus nerve was investigated in 20 truncally vagotomized subjects with pyloroplasty and 10 matched healthy controls. Subjects received GLP-1 (7-36 amide) or saline infusions during and after a standardized liquid mixed meal and a subsequent ad libitum meal. Despite no effect on appetite sensations, GLP-1 significantly reduced ad libitum food intake in the control group but had no effect in the vagotomized group. Gastric emptying was accelerated in vagotomized subjects and was decreased by GLP-1 in controls but not in vagotomized subjects. Postprandial glucose levels were reduced by the same percentage by GLP-1 in both groups. Peak postprandial GLP-1 levels were approximately fivefold higher in the vagotomized subjects. Insulin secretion was unaffected by exogenous GLP-1 in vagotomized subjects but was suppressed in controls. GLP-1 significantly reduced glucagon secretion in both groups, but levels were approximately twofold higher and were nonsuppressible in the early phase of the meal in vagotomized subjects. Our results demonstrate that vagotomy with pyloroplasty impairs the effects of exogenous GLP-1 on food intake, gastric emptying, and insulin and glucagon secretion, suggesting that intact vagal innervation may be important for GLP-1's actions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Human glucagon, European Pharmacopoeia (EP) Reference Standard