Merck
  • Lack of induction of direct protection or cross-protection in Staphylococcus aureus by sublethal concentrations of Origanum vulgare L. essential oil and carvacrol in a meat-based medium.
Lack of induction of direct protection or cross-protection in Staphylococcus aureus by sublethal concentrations of Origanum vulgare L. essential oil and carvacrol in a meat-based medium.
Archives of microbiology (2013-06-25)
Isabelle Silva da Luz, Nelson Justino Gomes Neto, Adassa Gama Tavares, Pollyana Campos Nunes, Marciane Magnani, Evandro Leite de Souza
ABSTRAKT
The capacity of Origanum vulgare L. essential oil (OVEO) and its majority compound, carvacrol (CAR), to induce direct tolerance and cross-tolerance in Staphylococcus aureus against high temperature (45 °C), lactic acid (pH 5.2) and NaCl (10 g/100 mL) was assessed. Overnight exposure of S. aureus to sublethal concentrations (1/2 MIC, 1/4 MIC) of either OVEO or CAR in meat broth revealed no induction of direct protection. S. aureus cells pre-adapted to OVEO or CAR showed no induction of cross-protection to high temperature, lactic acid or NaCl. Cells subjected to 24 h cycles of adaptation in increasing amounts (1/2 MIC to 2 × MIC) of OVEO or CAR showed no increase in direct tolerance. These results revealed a lack of induction of direct protection or cross-protection in S. aureus exposed to sublethal amounts of OVEO or CAR in meat-based broth, as determined by monitoring cell survival and growth behavior.