Merck
Efficient viral delivery system for unnatural amino acid mutagenesis in mammalian cells.
Proceedings of the National Academy of Sciences of the United States of America (2013-07-03)
Abhishek Chatterjee, Han Xiao, Michael Bollong, Hui-Wang Ai, Peter G Schultz
ABSTRAKT
Here we report the development of a baculovirus-based delivery system that enables the efficient incorporation of unnatural amino acids into proteins in mammalian cells. We have exploited the large cargo-capacity (>30 kb) and stability of the double-stranded DNA genome of baculovirus to deliver to a variety of cell types all of the components required to genetically incorporate novel amino acids. These include the engineered tRNA/aminoacyl-tRNA synthetase pair and the nonsense mutant of the target gene. Mammalian cell transduction efficiency of baculovirus was significantly improved by incorporating genetic elements from mammalian viruses. Two polyspecific tRNA/aminoacyl-tRNA synthetase pairs were inserted into this expression system, enabling the site-specific incorporation of a variety of unnatural amino acids with novel chemical and biological properties into proteins.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder