Merck
Temperature management in patients with acute neurological lesions: an Australian and New Zealand point prevalence study.
Critical care and resuscitation : journal of the Australasian Academy of Critical Care Medicine (2013-08-13)
Manoj K Saxena, Colman B Taylor, Naomi E Hammond, Paul J Young, Ian M Seppelt, Parisa Glass, John A Myburgh
ABSTRAKT
Given the scientific uncertainty of the efficacy and safety of normothermia (36.0°C to 37.5°C) on disability and death after acute neurological lesions, we sought to understand how temperature is managed in usual clinical care for this patient population in Australia and New Zealand. To describe temperature management in patients with acute neurological lesions. Prospective, observational, multicentre, single-day point-prevalence study. Observational data of usual practice were recorded for all patients with an intensive care admission diagnosis of acute neurological lesions and who were present in 33 intensive care units at 10:00 on the study day. Data were collected prospectively for the ensuing 24-hour period. Achieved temperature, interventions used to modify temperature and target temperature. There were 106 patients with acute neurological lesions (61% with either stroke or traumatic brain injury) with a mean APACHE (Acute Physiology and Chronic Health Evaluation) II score of 19.3 ± 7.4, age of 53.5 ± 19.0 years and median time from intensive care admission to data capture of 3 days (interquartile range, 1-9). A target temperature was specified in 24% of patients. Although paracetamol was commonly used (56%), it was infrequently used at the maximum licensed dose and there was no use recorded of non-steroidal anti-inflammatory drugs and cyclooxygenase-2 inhibitors. Physical cooling was used in 25% of patients and core temperature was measured in 32%. Measured temperature often exceeded 37.0°C (62% of readings), 37.5°C (43%) and 38.0°C (22%). Temperature readings above 37.5°C are common. Further cohort studies are required to validate these preliminary, exploratory findings.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder