Merck
[Vestibulo-protective properties of regulatory peptides].
Biulleten' eksperimental'noi biologii i meditsiny (1988-07-01)
Iu V Drozd, V V Iasnetsov, V S Shashkov
ABSTRAKT
Endogenous opioid peptides take an active part in the pathogenesis of motion sickness (MS). Some regulatory peptides (substances P, gamma and des-tyr-gamma-endorphins) manifest antiemetic properties, unlike opioid peptides and morphine. Thus, we have examined the vestibulo-protective properties of some regulatory peptides during simulation of MS in cats. Vestibulo-protective properties have been discovered in some peptides: sweet water hydra undecapeptide, substances P, gamma and des-tyr-gamma-endorphins. It has been suggested that regulatory peptides take an active part in the genesis of vestibulo-protective disorders during motion sickness.