Merck
The Pisa syndrome during maintenance antipsychotic therapy.
The American journal of psychiatry (1986-11-01)
N Guy, A Raps, M Assael
MATERIAŁY
Numer produktu
Marka
Opis produktu
Fluphenazine enantate, European Pharmacopoeia (EP) Reference Standard