Merck
Effects of inhibitors of diacylglycerol metabolism on protein kinase C-mediated responses in hepatocytes.
Biochimica et biophysica acta (1988-06-08)
T K Chataway, G J Barritt
ABSTRAKT
In hepatocytes pre-labelled with [3H]glycerol, compound R59022 (6-[2-(4-[(4-fluorophenyl)phenylmethylene]-1-piperidinyl)ethyl]-7- methyl-5H-thiazolo[3,2-alpha]pyrimidin-5-one) and 2-bromooctanoate each increased the amount of radioactivity in diacylglycerols. R59022 mimicked the actions of 12-O-tetradecanoylphorbol 13-acetate in completely abolishing the activation by adrenaline (but not that by vasopressin or glucagon) of glycogen phosphorylase a, and in decreasing the activity of glycogen synthetase. Exogenous dioctanoylglycerol caused a small inhibition of adrenaline-stimulated phosphorylase activity. The concentration of R59022 which gave half-maximal inhibition of adrenaline-stimulated phosphorylase activity was 15 microM. Maximal inhibition was observed within 2 min of addition of R59022. 2-Bromooctanoate activated phosphorylase by a process independent of changes in cyclic AMP and Ca2+, and decreased glycogen synthetase. It is concluded that in hepatocytes (i) diacylglycerols which accumulate as a result of the inhibition of diacylglycerol kinase by R59022 activate protein kinase C and (ii) 2-bromooctanoate increases diacylglycerols but also has other effects on hepatocyte metabolism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Bromooctanoic acid, 97%