Merck
Clinical applications of Ga-67 DFO-DAS-fibrinogen.
Radiation medicine (1987-09-01)
M Iio, T Ohtake, J Nishikawa, T Watanabe, H Yasuhara, S Ohashi, Y Takayama, Y Tada, M Shirakawa
ABSTRAKT
Ga-67 DFO-DAS-fibrinogen scanning was performed in 15 patients to detect fresh arterial thrombus. Seven patients were preoperative cases of aortic aneurysm and eight were postoperative cases of graft bypass. Scanning was performed on the first and second days after the injection. Nine of 13 grafts in eight cases were visualized by 67Ga-DFO-DAS-fibrinogen scan. Velour Dacron grafts were more strongly visualized compared with woven Dacron or Goatex. In three patients with aortic aneurysm in whom fresh thrombus was identified by operative findings, arterial thrombus was clearly visualized by 67Ga-DFO-DAS-fibrinogen scan, whereas in three other patients with aortic aneurysm, thrombus was not visualized, although it was identified by other examinations. In one case, a marked accumulation was observed in the abdominal aorta, although no arterial thrombus was found at operation. This was thought to be an accumulation in a blood pool because it decreased on the second day. We think it important to observe the relative changes of accumulation.