- Sensitivity to the weed killer DNA-nitralin and cross-sensitivity to dinitrochlorobenzene.
Sensitivity to the weed killer DNA-nitralin and cross-sensitivity to dinitrochlorobenzene.
Archives of dermatology (1983-04-01)
K Nishioka, C Asagami, M Kurata, H Fujita
PMID6220678
ABSTRAKT
A man, with a dermatitis acquired while working in a factory producing a weed killer, showed sensitivity to 4-methylsulfonyl 2,6-dinitro-N,N-dipropylaniline (DNA-nitralin) and its precursor, 4-chloro 3,-5-dinitrophenylmethyl sulfone (DNC), and cross-sensitivity to dinitrochlorobenzene (DNCB). Sensitization capacities of DNA-nitralin and DNC compared with DNCB, and cross-sensitizations among 11 dinitrobenzene derivatives, including DNA-nitralin, DNC, and DNCB, were studied in guinea pigs. We found that the order of potency was DNCB, DNC, and DNA-nitralin for the sensitization capacity, and that cross-sensitizations may occur among DNCB, DNC, DNA-nitralin, and dinitrofluorobenzene, in comparatively high incidence.