Merck
[3H]-dynorphin A (1-9): binding characteristics and degradation profile in brain homogenates.
Life sciences (1983-01-01)
L E Robson, M G Gillan, A T McKnight, H W Kosterlitz
ABSTRAKT
[3H]-Dynorphin A (1-9) is rapidly degraded in brain homogenates, even at 0 degree C. Although protection from degradation at O degree C, and to a lesser extent at 25 degrees C, was obtained with a combination of bestatin, L-leucyl-L-leucine, L-arginyl-L-arginine and captopril, these peptidase inhibitors had little effect on binding at 0 degree C. At the kappa-site, the maximum binding capacity of [3H]-dynorphin A (1-9) at 0 degree C was similar to that at 25 degrees C but it is suggested that, even after suppression of mu- and delta-binding, [3H]-dynorphin A (1-9) may bind to more than one site for which it has differing affinities at 0 degree C.