Merck
Allergic contact dermatitis from the preservative N-methylol-chloracetamide.
Contact dermatitis (1980-06-01)
S Agren, I Dahlquist, S Fregert, K Persson
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Chloro-N-(hydroxymethyl)acetamide, 98%