Merck
High-performance liquid chromatographic assay for prostaglandin E1 in various ointment vehicles. Separation and stability testing.
Journal of chromatography (1984-10-26)
K Yamamura, T Yotsuyanagi
ABSTRAKT
A high-performance liquid chromatographic method for the determination of prostaglandin E1 (PGE1) incorporated in white petrolatum, white ointment, hydrophilic petrolatum and Plastibase was investigated. The prostaglandin was separated from the oleaginous vehicles with n-hexane-aqueous acetonitrile. Most of white petrolatum, which is a principal component in the vehicles, remained in the n-hexane layer, and the recovery of the drug from any vehicle attained 100%. The method was applied to stability studies of PGE1 in white petrolatum and macrogol ointment. In which pure PGE1 and PGE1-alpha-cyclodextrin complex (PGE1-CD) were incorporated. The drug remained intact for up to 6 months when stored at 5 degrees C. At 25 and 40 degrees C, pure PGE1 was more stable than PGE1-CD and both PGE1 species were more stable in white petrolatum than in macrogol ointment.