Merck
Agonist-antagonist actions and stereoselectivity of optical pairs of some N-substituted alpha-N-normetazocines.
Journal of pharmaceutical sciences (1984-12-01)
M D Aceto, E L May, L S Harris, A E Jacobson, H Awaya
ABSTRAKT
alpha-(-)-, (+)-, and (+/-)-N-4-Methylpentyl-, (-)- and (+)-N-cis-3-chloroallyl-, and (-)- and (+)-N-propynyl-N-normetazocine (I, II, and III, respectively) have been prepared from alpha-(-)-, (+)-, and (+/-)-N-normetazocine (IV) and tested for antinociceptive activity in mice and in morphine-dependent rhesus monkeys. The results obtained with Ia and b somewhat resemble the results obtained with the corresponding phenazocine isomers, whereas those observed with IIa and b more nearly correspond with those reported for (+)- and (-)-N-allyl-N-normetazocine (SKF 10,047; NANM). The propynyl isomers (IIIa and b) display profiles of activity more closely like the corresponding isomers of metazocine. Evidence is considered which suggests that some of the (+)-isomers merit closer scrutiny in animal and human studies.