Merck
Indolealkylamine and phenalkylamine hallucinogens. Effect of alpha-methyl and N-methyl substituents on behavioral activity.
Biochemical pharmacology (1983-04-01)
R A Glennon, R Young, J M Jacyno
ABSTRAKT
Animals (rats), trained to discriminate the hallucinogenic agent 1-(2,5-dimethoxy-4-methylphenyl)-2-aminopropane (DOM) from saline in a two-lever operant procedure, were challenged with various doses of several indolealkylamine and phenalkylamine derivatives. In both series, the alpha-methyl analogs were found to be more active than either their N-methyl or alpha-demethyl counterparts. Furthermore, when the activities of the optical isomers of DOM were compared with the activities of S-(+) and R-(-)-alpha-methyltryptamine (alpha-MeT), it was found that the more potent isomer of alpha-MeT (i.e. S) possessed the opposite absolute configuration of the more potent isomer of DOM (i.e. R). With respect to the mechanism of action of these agents, these findings are not inconsistent with a common site hypothesis.