Merck
An improved method for the histochemical localization of glucose-6-phoshate dehydrogenase in animal and plant tissues.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1977-02-01)
D S Negi, R J Stephens
PMID839060
ABSTRAKT
An improved procedure is reported for the histochemical localization of glucose-6-phosphate dehydrogenase in fresh-frozen sections of animal and plant tissues. Used in this procedure are an incubation medium and an inert high-molecular-weight polyvinyl alcohol as a stabilizer to check enzyme diffusion. The incubation medium consists of 10 mg each of nicotinamide adenine dinucleotide phosphate and nitroblue tetrazolium chloride and 60 mg of glucose-6-phosphate per 20 ml of 22% polyvinyl alcohol solution in 0.2 M Tris-maleate buffer, pH 7.2. Use of this medium has given consistent results with the tissues tested. Because the use of an electron-transfer agent in the incubation medium often resulted in enzyme diffusion, its use in glucose-6-phosphate dehydrogenase localization is considered undersirable. Incubation time of 15 to 30 min for animal tissue and 20 to 45 min for plant tissue of 37degreesC was found to be optimal for producing the characteristic blue formazan at the reaction sites.