Merck
Serum-sulfated lithocholate as an indicator of cholestasis during parenteral nutrition in infants and children.
JPEN. Journal of parenteral and enteral nutrition (1982-01-01)
M K Farrell, W F Balistreri, F J Suchy