- Novel carbon-carbon bond cleavage reaction of penem antibiotic by class C beta-lactamases.
Novel carbon-carbon bond cleavage reaction of penem antibiotic by class C beta-lactamases.
The Journal of antibiotics (1994-08-01)
R Tanaka, K Namikawa, T Nakatsuka, H Adachi, T Yoshida, O Sugita, M Ishiguro
PMID7928684
MATERIAŁY