- Kinetics of inhibition of MAO-B by N-(2-aminoethyl)-p-chlorobenzamide (Ro 16-6491) and analogues.
Kinetics of inhibition of MAO-B by N-(2-aminoethyl)-p-chlorobenzamide (Ro 16-6491) and analogues.
Journal of neural transmission. Supplementum (1994-01-01)
E L Mullan, C H Williams
PMID7931243
ABSTRAKT
Ro 16-6491 is known to be a potent reversible inhibitor of human brain MAO-B. This compound and several analogues were tested for their effect on bovine liver MAO-B. It was found that in compounds where the amide bond of Ro 16-6491 was replaced by an ester bond an increase in potency of the order of 150-200 times was obtained.