- [Antioxidant activity of a new derivative of 4-hydrazinoquinazoline in experimental cerebral hypoxia].
[Antioxidant activity of a new derivative of 4-hydrazinoquinazoline in experimental cerebral hypoxia].
Ukrainskii biokhimicheskii zhurnal (1978) (1993-05-01)
V V Dunaev, I F Belenichev, S I Kovalenko, I A Mazur, R S Siniak, I N Bashkin, V S Tishkin
PMID8291133
ABSTRAKT
It is established that KS-79 compound possesses the high level of antioxidant activity and realizes its antioxidant effect at initial stages of free-radical hydrogen peroxide oxidation of lipids. Due to this ability it differs from the oxidants of direct-type action.